COM448634519 Combi-Blocks, Inc.

3-(Tetrahydropyran-4-yloxy)-5-trifluoromethylphenylboronic acid, pinacol ester

  • MDL number MFCD18434454
  • Cas Number : 2096341-24-9
  • Molecular Weight : 372.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6701
Availability details for
Quantity
  • Availability
  • Add to Cart