COM448634524 Combi-Blocks, Inc.

4-(2-Hydroxy-3-pyrrolidinopropoxy)phenylboronic acid, pinacol ester

  • MDL number MFCD18087746
  • Cas Number : 2096334-38-0
  • Molecular Weight : 347.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6713
Availability details for
Quantity
  • Availability
  • Add to Cart