COM448634528 Combi-Blocks, Inc.

2-Chloro-4-(N-cyclopentylaminomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18311893
  • Cas Number : 2096337-34-5
  • Molecular Weight : 335.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6726
Availability details for
Quantity
  • Availability
  • Add to Cart