COM448634537 Combi-Blocks, Inc.

2-(Diisopropylaminomethyl)-5-fluorophenylboronic acid pinacol ester

  • MDL number MFCD18434460
  • Cas Number : 2096332-47-5
  • Molecular Weight : 335.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6737
Availability details for
Quantity
  • Availability
  • Add to Cart