COM448634538 Combi-Blocks, Inc.

3-Methylpyridazine-5-boronic acid, pinacol ester

  • MDL number MFCD18311901
  • Cas Number : 1350543-95-1
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6750
Availability details for
Quantity
  • Availability
  • Add to Cart