COM448634540 Combi-Blocks, Inc.

4-[(Tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]morpholine

  • MDL number MFCD24448819
  • Cas Number : 70558-05-3
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6785
Availability details for
Quantity
  • Availability
  • Add to Cart