COM448634551 Combi-Blocks, Inc.

3-Chloro-4-(N-cyclohexylaminomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18837626
  • Cas Number : 2096329-99-4
  • Molecular Weight : 349.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6806
Availability details for
Quantity
  • Availability
  • Add to Cart