COM448634552 Combi-Blocks, Inc.

3-Chloro-4-(piperidinoaminomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18434466
  • Cas Number : 2096341-25-0
  • Molecular Weight : 335.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6807
Availability details for
Quantity
  • Availability
  • Add to Cart