COM448634553 Combi-Blocks, Inc.

2-Chloro-4-hydroxymethylphenylboronic acid, pinacol ester

  • MDL number MFCD18434467
  • Cas Number : 1485110-17-5
  • Molecular Weight : 268.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6808
Availability details for
Quantity
  • Availability
  • Add to Cart