COM448634555 Combi-Blocks, Inc.

4-(N-Cbz-N-Methylamino)phenylboronic acid, pinacol ester

  • MDL number MFCD18311903
  • Cas Number : 2096334-55-1
  • Molecular Weight : 367.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6821
Availability details for
Quantity
  • Availability
  • Add to Cart