COM448634557 Combi-Blocks, Inc.

3-Cyano-2-(N,N-dimethylamino)pyridine-5-boronic acid, pinacol ester

  • MDL number MFCD16995282
  • Cas Number : 1346809-50-4
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6830
Availability details for
Quantity
  • Availability
  • Add to Cart