COM448634559 Combi-Blocks, Inc.

3-Cyano-2-morpholinopyridine-5-boronic acid, pinacol ester

  • MDL number MFCD16996767
  • Cas Number : 1356068-62-6
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6832
Availability details for
Quantity
  • Availability
  • Add to Cart