COM448634561 Combi-Blocks, Inc.

2,3-Difluoro-4-(methoxycarbonyl)phenylboronic acid pinacol ester

  • MDL number MFCD27981324
  • Cas Number : 1621256-23-2
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6843
Availability details for
Quantity
  • Availability
  • Add to Cart