COM448634566 Combi-Blocks, Inc.

4-Bromomethyl-2-fluorophenylboronic acid pinacol ester

  • MDL number MFCD18434471
  • Cas Number : 1029439-49-3
  • Molecular Weight : 315.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6853
Availability details for
Quantity
  • Availability
  • Add to Cart