COM448634570 Combi-Blocks, Inc.

2,4-Dichloro-5-(3-pyridylmethoxy)phenylboronic aicd pinacol ester

  • MDL number MFCD18837627
  • Cas Number : 2377607-60-6
  • Molecular Weight : 380.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6877
Availability details for
Quantity
  • Availability
  • Add to Cart