COM448634572 Combi-Blocks, Inc.

3-Chloro-5-pyrrolidinophenylboronic acid, pinacol ester

  • MDL number MFCD18783182
  • Cas Number : 2096331-84-7
  • Molecular Weight : 307.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6882
Availability details for
Quantity
  • Availability
  • Add to Cart