COM448634573 Combi-Blocks, Inc.

2-(t-Butoxycarbonyl)-6-chloropyridine-4-boronic acid, pinacol ester

  • MDL number MFCD18783183
  • Cas Number : 2096341-89-6
  • Molecular Weight : 339.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6887
Availability details for
Quantity
  • Availability
  • Add to Cart