COM448634575 Combi-Blocks, Inc.

3-Chloro-2-(4-methylpiperazino)pyridine-5-boronic acid, pinacol ester

  • MDL number MFCD16996502
  • Cas Number : 2096334-60-8
  • Molecular Weight : 337.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6897
Availability details for
Quantity
  • Availability
  • Add to Cart