COM448634580 Combi-Blocks, Inc.

5-(4-BOC-piperazinomethyl)-2-chlorophenylboronic acid, pinacol ester

  • MDL number MFCD19237217
  • Cas Number : 2096334-64-2
  • Molecular Weight : 436.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6959
Availability details for
Quantity
  • Availability
  • Add to Cart