COM448634582 Combi-Blocks, Inc.

2-Chloro-5-(N,N-diethylaminomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18783188
  • Cas Number : 2096341-29-4
  • Molecular Weight : 323.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6964
Availability details for
Quantity
  • Availability
  • Add to Cart