COM448634583 Combi-Blocks, Inc.

5-(4-Methylpiperazinomethyl)-2-chlorophenylboronic acid, pinacol ester

  • MDL number MFCD19684131
  • Cas Number : 2096336-69-3
  • Molecular Weight : 350.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6965
Availability details for
Quantity
  • Availability
  • Add to Cart