COM448634584 Combi-Blocks, Inc.

2-Chloro-5-(pyrrolidinomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18783189
  • Cas Number : 2096333-77-4
  • Molecular Weight : 321.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6966
Availability details for
Quantity
  • Availability
  • Add to Cart