COM448634585 Combi-Blocks, Inc.

2-Chloro-5-(piperidinomethyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18783190
  • Cas Number : 2096334-70-0
  • Molecular Weight : 335.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6967
Availability details for
Quantity
  • Availability
  • Add to Cart