COM448634586 Combi-Blocks, Inc.

5-(N-t-Butylaminomethyl)-2-chlorophenylboronic acid, pinacol ester

  • MDL number MFCD18783191
  • Cas Number : 2096333-91-2
  • Molecular Weight : 323.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6968
Availability details for
Quantity
  • Availability
  • Add to Cart