COM448634593 Combi-Blocks, Inc.

3,5-Dimethyl-2-fluorophenylboronic acid, pinacol ester

  • MDL number MFCD18914525
  • Cas Number : 2096341-90-9
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6994
Availability details for
Quantity
  • Availability
  • Add to Cart