COM448634594 Combi-Blocks, Inc.

5-Bromo-2-fluoro-4-(methoxycarbonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18837629
  • Cas Number : 2096331-46-1
  • Molecular Weight : 359.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6999
Availability details for
Quantity
  • Availability
  • Add to Cart