COM448634596 Combi-Blocks, Inc.

1-Methyl-1H-pyrrole-2,4-diboronic acid, pinacol ester

  • MDL number MFCD18914526
  • Cas Number : 1278579-60-4
  • Molecular Weight : 333.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7023
Availability details for
Quantity
  • Availability
  • Add to Cart