COM448634599 Combi-Blocks, Inc.

4-Chloro-3-(methoxycarbonyl)-5-methylphenylboronic acid, pinacol ester

  • MDL number MFCD18914528
  • Cas Number : 2096330-10-6
  • Molecular Weight : 310.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7027
Availability details for
Quantity
  • Availability
  • Add to Cart