COM448634602 Combi-Blocks, Inc.

3,4-Dichloro-5-methoxyphenylphenylboronic acid, pinacol ester

  • MDL number MFCD19237221
  • Cas Number : 1701449-89-9
  • Molecular Weight : 303.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7038
Availability details for
Quantity
  • Availability
  • Add to Cart