COM448634604 Combi-Blocks, Inc.

2-Fluoro-3-methyl-5-(methoxycarbonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD19237222
  • Cas Number : 2096330-13-9
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7040
Availability details for
Quantity
  • Availability
  • Add to Cart