COM448634606 Combi-Blocks, Inc.

4-Acetyl-2-methylphenylboronic acid pinacol ester

  • MDL number MFCD18732916
  • Cas Number : 1321848-43-4
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-7048
Availability details for
Quantity
  • Availability
  • Add to Cart