COM448634610 Combi-Blocks, Inc.

Perfluorophenylboronic acid, pinacol ester

  • MDL number MFCD12405352
  • Cas Number : 325142-81-2
  • Molecular Weight : 294.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8007
Availability details for
Quantity
  • Availability
  • Add to Cart