COM448634615 Combi-Blocks, Inc.

3-Bromo-5-isopropoxyphenylboronic acid, pinacol ester

  • MDL number MFCD12546586
  • Cas Number : 1218790-34-1
  • Molecular Weight : 341.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8075
Availability details for
Quantity
  • Availability
  • Add to Cart