COM448634628 Combi-Blocks, Inc.

2-Chloro-3-fluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD07787387
  • Cas Number : 1029654-43-0
  • Molecular Weight : 257.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8244

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8244
Availability details for
Quantity
  • Availability
  • Add to Cart