COM448634630 Combi-Blocks, Inc.

Isobutylboronic acid pinacol ester

  • MDL number MFCD17015837
  • Cas Number : 67562-20-3
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8309
Availability details for
Quantity
  • Availability
  • Add to Cart