COM448634636 Combi-Blocks, Inc.

3-Methanesulfinylphenylboronic acid, pinacol ester

  • MDL number MFCD18733605
  • Cas Number : 1416367-04-8
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8368
Availability details for
Quantity
  • Availability
  • Add to Cart