COM448634639 Combi-Blocks, Inc.

5-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

  • MDL number MFCD22418302
  • Cas Number : 1021868-08-5
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8379

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8379
Availability details for
Quantity
  • Availability
  • Add to Cart