COM448634642 Combi-Blocks, Inc.

3-(Pyrrilidyl)phenylboronic acid pinacol ester

  • MDL number MFCD08271900
  • Cas Number : 857283-63-7
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8384
Availability details for
Quantity
  • Availability
  • Add to Cart