COM448634669 Combi-Blocks, Inc.

2-Bromo-5-methoxypyridine-4-boronic acid, pinacol ester

  • MDL number MFCD22421547
  • Cas Number : 2096334-49-3
  • Molecular Weight : 314.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8532
Availability details for
Quantity
  • Availability
  • Add to Cart