COM448634670 Combi-Blocks, Inc.

4,5-Dimethoxy-2-(methoxycarbonyl)phenylboronic acid pinacol ester

  • MDL number MFCD16036165
  • Cas Number : 1201566-80-4
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8533
Availability details for
Quantity
  • Availability
  • Add to Cart