COM448634688 Combi-Blocks, Inc.

3,3-Diethoxy-1-propylboronic acid pinacol ester

  • MDL number MFCD03788723
  • Cas Number : 165904-27-8
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8612
Availability details for
Quantity
  • Availability
  • Add to Cart