COM448634720 Combi-Blocks, Inc.

N-[2-Hydroxyethyl]benzamide-3-boronic acid, pinacol ester

  • MDL number MFCD06795647
  • Cas Number : 943911-66-8
  • Molecular Weight : 291.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8664
Availability details for
Quantity
  • Availability
  • Add to Cart