COM448634735 Combi-Blocks, Inc.

2-Cyanomethoxyphenylboronic acid, pinacol ester

  • MDL number MFCD06795686
  • Cas Number : 936250-19-0
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8681
Availability details for
Quantity
  • Availability
  • Add to Cart