COM448634783 Combi-Blocks, Inc.

2-[4-(N-Boc)piperazin-1-yl]phenylboronic acid pinacol ester

  • MDL number MFCD08669542
  • Cas Number : 1073354-59-2
  • Molecular Weight : 388.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8746
Availability details for
Quantity
  • Availability
  • Add to Cart