COM448634824 Combi-Blocks, Inc.

3-[3-(Ethoxycarbonyl)piperidine-1-carbonyl]phenylboronic acid pinacol ester

  • MDL number MFCD09266178
  • Cas Number : 1073355-04-0
  • Molecular Weight : 387.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8798
Availability details for
Quantity
  • Availability
  • Add to Cart