COM448634840 Combi-Blocks, Inc.

3-Acetoxy-4-methoxycarbonylphenylboronic acid, pinacol ester

  • MDL number MFCD09864187
  • Cas Number : 1073355-18-6
  • Molecular Weight : 320.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8816
Availability details for
Quantity
  • Availability
  • Add to Cart