COM448634849 Combi-Blocks, Inc.

2-Phenyl-1-propylboronic acid pinacol ester

  • MDL number MFCD09953475
  • Cas Number : 280559-30-0
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8828
Availability details for
Quantity
  • Availability
  • Add to Cart