COM448634851 Combi-Blocks, Inc.

3-(2-Hydroxyphenyl)propylboronic acid, pinacol ester

  • MDL number MFCD09953481
  • Cas Number : 1073355-25-5
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8833
Availability details for
Quantity
  • Availability
  • Add to Cart