COM448634863 Combi-Blocks, Inc.

4-(BOC-Methylamino)phenylboronic acid, pinacol ester

  • MDL number MFCD03490498
  • Cas Number : 916587-44-5
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8852
Availability details for
Quantity
  • Availability
  • Add to Cart