COM448634880 Combi-Blocks, Inc.

3-(3-(Dimethylamino)propylcarbamoyl)phenylboronic acid, pinacol ester

  • MDL number MFCD05865085
  • Cas Number : 936250-16-7
  • Molecular Weight : 332.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8870
Availability details for
Quantity
  • Availability
  • Add to Cart